Assay Details
Target Gene Details
Entrez Gene ID: | 11605 |
Gene Name: | galactosidase, alpha |
Gene Aliases: |
Ags |
Location: |
Chr.X:134588169-134601005 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gla | NM_013463.2 | 6 | 935 | NP_038491.2 |
| AK040081.1 | 6 | 867 | BAC30508.1 | |
| AK054547.1 | 6 | 887 | BAC35819.1 | |
| AK145011.1 | 6 | 877 | ||
| AK164231.1 | 6 | 868 | BAE37692.1 | |
| AK166204.1 | 6 | 868 | BAE38627.1 | |
| AK166245.1 | 6 | 884 | BAE38657.1 | |
| BC009021.1 | 6 | 888 | AAH09021.1 | |
| U34071.1 | 6 | 885 | AAA96749.1 | |
| U50716.1 | 6 | 873 | AAC52584.1 |
More Information
Set Membership: |
|

Genomic Map