Assay Details
Target Gene Details
Entrez Gene ID: | 13058 |
Gene Name: | cytochrome b-245, beta polypeptide |
Gene Aliases: |
C88302, CGD91-phox, Cgd, Cyd, Nox2, gp91-1, gp91phox |
Location: |
Chr.X:9435254-9487766 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cybb | NM_007807.5 | 10 | 1318 | NP_031833.3 |
| AK033570.1 | 10 | 1263 | ||
| AK053920.1 | 10 | 1262 | BAC35593.1 | |
| AK152257.1 | 10 | 1289 | BAE31076.1 | |
| AK153071.1 | 3 | 428 | BAE31697.1 | |
| AK153154.1 | 10 | 1289 | BAE31764.1 | |
| AK153259.1 | 10 | 1289 | BAE31850.1 | |
| AK154126.1 | 10 | 1318 | BAE32391.1 | |
| AK160031.1 | 5 | 720 | BAE35574.1 | |
| AK171526.1 | 10 | 1304 | BAE42507.1 | |
| BC071229.1 | 10 | 1273 | AAH71229.1 | |
| FJ168469.1 | 10 | 1307 | ||
| U43384.1 | 10 | 1244 | AAB05997.1 |
More Information
Set Membership: |
|

Genomic Map