Assay Details
Target Gene Details
Entrez Gene ID: | 73809 |
Gene Name: | spermidine/spermine N1-acetyl transferase-like 1 |
Gene Aliases: |
4930404K22Rik |
Location: |
Chr.X:112383207-112406779 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Satl1 | NM_028655.1 | NP_082931.1 | ||
| AK015086.1 | BAB29707.1 | |||
| BC138009.1 | ||||
| BC138011.1 |
More Information
Set Membership: |
|

Genomic Map