Assay Details
Target Gene Details
Entrez Gene ID: | 236852 |
Gene Name: | melanoma antigen family A, 10 |
Gene Aliases: |
- |
Location: |
Chr.X:72381870-72386858 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Magea10 | NM_001085506.1 | NP_001078975.1 | ||
| AK136064.1 |
More Information
Set Membership: |
|

Genomic Map