Assay Details
Target Gene Details
Entrez Gene ID: | 54645 |
Gene Name: | GRIP1 associated protein 1 |
Gene Aliases: |
AI854681, DXImx47e, GRASP-1, Sfc10, mKIAA1167 |
Location: |
Chr.X:7789961-7820567 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gripap1 | NM_001290455.1 | 16 | 1431 | NP_001277384.1 |
NM_207670.2 | 15 | 1338 | NP_997553.1 | |
AK037997.1 | 9 | 1793 | ||
AK156165.1 | 15 | 1339 | BAE33608.1 | |
AK158098.1 | 12 | 1136 | BAE34355.1 | |
AK172590.1 | 13 | 1204 | BAE43083.1 | |
AK172623.1 | 15 | 1339 | BAE43101.1 | |
AK172699.1 | 15 | 1339 | BAE43133.1 | |
AK173119.1 | 3 | 354 | BAD32397.1 | |
AW342118.1 | 15 | 1313 | ||
BC138523.1 | 16 | 1427 | ||
BC145310.1 | 16 | 1427 |
