Assay Details
Target Gene Details
Entrez Gene ID: | 54645 |
Gene Name: | GRIP1 associated protein 1 |
Gene Aliases: |
AI854681, DXImx47e, GRASP-1, Sfc10, mKIAA1167 |
Location: |
Chr.X:7789961-7820567 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gripap1 | NM_001290455.1 | 22 | 2046 | NP_001277384.1 |
NM_207670.2 | 21 | 1953 | NP_997553.1 | |
AK156165.1 | 21 | 1954 | BAE33608.1 | |
AK172623.1 | 21 | 1954 | BAE43101.1 | |
AK172699.1 | 21 | 1954 | BAE43133.1 | |
AK173119.1 | 9 | 969 | BAD32397.1 | |
AK179078.1 | ||||
AW342118.1 | 21 | 1928 | ||
BC138523.1 | 22 | 2042 | ||
BC145310.1 | 22 | 2042 |
