Assay Details
Target Gene Details
Entrez Gene ID: | 54645 |
Gene Name: | GRIP1 associated protein 1 |
Gene Aliases: |
AI854681, DXImx47e, GRASP-1, Sfc10, mKIAA1167 |
Location: |
Chr.X:7789961-7820567 on Build GRCm38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gripap1 | NM_001290455.1 | 25 | 2378 | NP_001277384.1 |
| NM_207670.2 | 24 | 2285 | NP_997553.1 | |
| AK156165.1 | 24 | 2286 | BAE33608.1 | |
| AK160472.1 | 1 | 327 | ||
| AK172623.1 | 24 | 2286 | BAE43101.1 | |
| AK172699.1 | 24 | 2286 | BAE43133.1 | |
| AK173119.1 | 11 | 2078 | BAD32397.1 | |
| AW342118.1 | 24 | 2260 | ||
| BC138523.1 | 25 | 2374 | ||
| BC145310.1 | 25 | 2374 | ||
| BY708466.1 | 1 | 327 |

Genomic Map