Assay Details
Target Gene Details
Entrez Gene ID: | 209268 |
Gene Name: | immunoglobulin superfamily, member 1 |
Gene Aliases: |
5330413N23, 5530402E03, AI747649, InhBP/p120, mKIAA0364 |
Location: |
Chr.X:49782536-49797906 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Igsf1 | NM_177591.4 | 19 | 3943 | NP_808259.2 |
| NM_183336.2 | 10 | 2197 | NP_899179.1 | |
| AK030701.1 | 10 | 2199 | ||
| AK076427.1 | 10 | 2190 | BAC36338.1 | |
| AK129126.1 | 17 | 3066 | BAC97936.1 | |
| AY227771.1 | 19 | 3937 | AAP57079.1 | |
| AY227774.1 | 10 | 2192 | AAP57082.1 | |
| BC119264.1 | 10 | 2160 | ||
| BC119266.1 | 10 | 2160 |
More Information
Set Membership: |
|

Genomic Map