Assay Details
Target Gene Details
Entrez Gene ID: | 56258 |
Gene Name: | heterogeneous nuclear ribonucleoprotein H2 |
Gene Aliases: |
DXHXS1271E, Ftp-3, Ftp3, H', HNRNP, Hnrph2 |
Location: |
Chr.X:134601184-134607060 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hnrnph2 | NM_019868.3 | NP_063921.1 | ||
| AK077716.1 | BAC36976.1 | |||
| AK141861.1 | BAE24862.1 | |||
| AK164115.1 | BAE37634.1 | |||
| AK168116.1 | BAE40087.1 | |||
| AK169104.1 | BAE40886.1 | |||
| BC005461.1 | AAH05461.1 |
More Information
Set Membership: |
|

Genomic Map