Assay Details
Target Gene Details
Entrez Gene ID: | 109731 |
Gene Name: | monoamine oxidase B |
Gene Aliases: |
6330414K01Rik, MAO-B |
Location: |
Chr.X:16709281-16817366 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Maob | NM_172778.2 | 8 | 944 | NP_766366.2 |
| AK031833.1 | 8 | 928 | BAC27571.1 | |
| AK054050.1 | 8 | 943 | BAC35634.1 | |
| BC113182.1 | 8 | 933 | ||
| BC113788.1 | 8 | 933 |
More Information
Set Membership: |
|

Genomic Map