Assay Details
Target Gene Details
Entrez Gene ID: | 16728 |
Gene Name: | L1 cell adhesion molecule |
Gene Aliases: |
CD171, L1, N-CAM-L1, NCAM-L1 |
Location: |
Chr.X:73853780-73880841 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
L1cam | NM_008478.3 | 13 | 1715 | NP_032504.3 |
AJ627046.1 | 5 | 660 | CAF25314.1 | |
BC056988.1 | 13 | 1640 | AAH56988.1 | |
X12875.1 | 12 | 1462 | CAA31368.1 |
