Assay Details
Target Gene Details
Entrez Gene ID: | 72459 |
Gene Name: | HIV TAT specific factor 1 |
Gene Aliases: |
1600023H17Rik, 2600017A12Rik, 2700077B20Rik, TAT-SF1 |
Location: |
Chr.X:57053570-57067183 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Htatsf1 | NM_028242.2 | 6 | 910 | NP_082518.1 |
| NM_029371.1 | 5 | 834 | NP_083647.1 | |
| AK011234.1 | 5 | 781 | BAB27483.1 | |
| AK049495.1 | 5 | 766 | BAC33777.1 | |
| BC037711.1 | 6 | 875 | AAH37711.1 |
More Information
Set Membership: |
|

Genomic Map