Assay Details
Target Gene Details
| Entrez Gene ID: | 16728 | 
| Gene Name: | L1 cell adhesion molecule | 
| Gene Aliases: | CD171, L1, N-CAM-L1, NCAM-L1 | 
| Location: | Chr.X:73853780-73880841 on Build GRCm38 | 
| Assay Gene Location: | Overlaps Intron 23 - Exon 24 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| L1cam | NM_008478.3 | NP_032504.3 | ||
| BC056988.1 | AAH56988.1 | |||
| X12875.1 | CAA31368.1 | 
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top Intragenic
							Intragenic