Assay Details
Target Gene Details
Entrez Gene ID: | 16728 |
Gene Name: | L1 cell adhesion molecule |
Gene Aliases: |
CD171, L1, N-CAM-L1, NCAM-L1 |
Location: |
Chr.X:73853780-73880841 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
L1cam | NM_008478.3 | NP_032504.3 | ||
BC056988.1 | AAH56988.1 | |||
X12875.1 | CAA31368.1 |
