Assay Details
Target Gene Details
Entrez Gene ID: | 15185 |
Gene Name: | histone deacetylase 6 |
Gene Aliases: |
Hd6, Hdac5, Sfc6, mHDA2 |
Location: |
Chr.X:7930120-7947889 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hdac6 | NM_001130416.1 | NP_001123888.1 | ||
NM_010413.3 | NP_034543.3 | |||
AF006603.2 | AAD09835.2 | |||
AK148092.1 | ||||
AK148144.1 | BAE28372.1 | |||
AK154099.1 | BAE32376.1 | |||
AK155545.1 | 13 | 1285 | ||
AK155546.1 | ||||
BC041105.1 | AAH41105.1 |
