Assay Details
Target Gene Details
Entrez Gene ID: | 15185 |
Gene Name: | histone deacetylase 6 |
Gene Aliases: |
Hd6, Hdac5, Sfc6, mHDA2 |
Location: |
Chr.X:7930120-7947889 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hdac6 | NM_001130416.1 | 17 | 1648 | NP_001123888.1 |
NM_010413.3 | 17 | 1723 | NP_034543.3 | |
AF006603.2 | 16 | 1408 | AAD09835.2 | |
AK148092.1 | 17 | 1569 | ||
AK148144.1 | 17 | 1539 | BAE28372.1 | |
AK154099.1 | 17 | 1587 | BAE32376.1 | |
AK155545.1 | 15 | 2621 | ||
AK155546.1 | 17 | 1587 | ||
AK183013.1 | 3 | 181 | ||
BC041105.1 | 17 | 1580 | AAH41105.1 |
