Assay Details
Target Gene Details
Entrez Gene ID: | 26549 |
Gene Name: | integrin beta 1 binding protein 2 |
Gene Aliases: |
Chordc3 |
Location: |
Chr.X:101448690-101466122 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Itgb1bp2 | NM_013712.2 | NP_038740.1 | ||
| AF140691.1 | AAF01677.1 | |||
| AK003906.1 | BAB23069.1 | |||
| BC030035.1 | AAH30035.1 |
More Information
Set Membership: |
|

Genomic Map