Assay Details
Target Gene Details
Entrez Gene ID: | 54405 |
Gene Name: | NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 1 |
Gene Aliases: |
1810049F12Rik, MWFE |
Location: |
Chr.X:37187588-37191238 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndufa1 | NM_019443.2 | NP_062316.1 | ||
| AI844651.1 | ||||
| AK007840.1 | BAB25297.1 | |||
| BC024364.1 | AAH24364.1 | |||
| CF383819.1 | ||||
| CT010398.1 | CAJ18604.1 | |||
| Y07708.1 | CAA68977.1 |
More Information
Set Membership: |
|

Genomic Map