Assay Details
Target Gene Details
Entrez Gene ID: | 104130 |
Gene Name: | NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 11 |
Gene Aliases: |
D5Bwg0566e, D5Bwg0577e, NP15.6, Np15 |
Location: |
Chr.X:20615326-20617564 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndufb11 | NM_019435.4 | NP_062308.2 | ||
| AK028121.1 | BAC25760.1 | |||
| BC027265.1 | AAH27265.1 | |||
| BC096047.1 | AAH96047.1 | |||
| DV654320.1 | ||||
| Y08702.1 | CAA69961.1 |
More Information
Set Membership: |
|

Genomic Map