Assay Details
Target Gene Details
Entrez Gene ID: | 102857 |
Gene Name: | solute carrier family 6 (neurotransmitter transporter, creatine), member 8 |
Gene Aliases: |
AA589632, CRT, CRTR, CT1, CTR5, Creat |
Location: |
Chr.X:73673133-73682502 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 11 - Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc6a8 | NM_001142809.1 | NP_001136281.1 | ||
| NM_001142810.1 | NP_001136282.1 | |||
| NM_133987.2 | NP_598748.1 | |||
| AB077327.1 | BAC11857.1 | |||
| AF459435.1 | AAL66354.1 | |||
| AJ748650.1 | CAG38684.1 | |||
| AK201382.1 | ||||
| BC024444.1 | AAH24444.1 | |||
| BC049801.1 | AAH49801.1 | |||
| BC141067.1 | ||||
| BC145319.1 | ||||
| EU523148.1 | ||||
| KC994641.1 |
More Information
Set Membership: |
|

Genomic Map