Assay Details
Target Gene Details
Entrez Gene ID: | 15185 |
Gene Name: | histone deacetylase 6 |
Gene Aliases: |
Hd6, Hdac5, Sfc6, mHDA2 |
Location: |
Chr.X:7930120-7947889 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hdac6 | NM_001130416.1 | 20 | 2131 | NP_001123888.1 |
NM_010413.3 | 20 | 2206 | NP_034543.3 | |
AF006603.2 | 19 | 1891 | AAD09835.2 | |
AK148092.1 | 20 | 2052 | ||
AK148144.1 | 20 | 2022 | BAE28372.1 | |
AK154099.1 | 20 | 2070 | BAE32376.1 | |
AK155545.1 | 18 | 3104 | ||
AK155546.1 | 20 | 2070 | ||
AK177646.1 | 2 | 103 | ||
AK184333.1 | 2 | 103 | ||
BC041105.1 | 20 | 2063 | AAH41105.1 |
