Assay Details
Target Gene Details
Entrez Gene ID: | 15185 |
Gene Name: | histone deacetylase 6 |
Gene Aliases: |
Hd6, Hdac5, Sfc6, mHDA2 |
Location: |
Chr.X:7930120-7947889 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hdac6 | NM_001130416.1 | 22 | 2322 | NP_001123888.1 |
NM_010413.3 | 22 | 2397 | NP_034543.3 | |
AF006603.2 | 21 | 2082 | AAD09835.2 | |
AK148092.1 | 22 | 2243 | ||
AK148144.1 | 22 | 2213 | BAE28372.1 | |
AK154099.1 | 22 | 2261 | BAE32376.1 | |
AK155545.1 | 20 | 3295 | ||
AK155546.1 | 22 | 2261 | ||
BC041105.1 | 22 | 2254 | AAH41105.1 |
