Assay Details
Target Gene Details
Entrez Gene ID: | 22290 |
Gene Name: | ubiquitously transcribed tetratricopeptide repeat gene, Y chromosome |
Gene Aliases: |
Hydb, mKIAA4057 |
Location: |
Chr.Y:1085187-1245754 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 19 - Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Uty | NM_009484.2 | NP_033510.2 | ||
| AF057367.1 | AAC67385.1 | |||
| AK142512.1 | BAE25094.1 | |||
| AK162410.1 | BAE36901.1 | |||
| AK164917.1 | BAE37965.1 | |||
| AK220463.1 | BAD90490.1 |
More Information
Set Membership: |
|

Genomic Map