Assay Details
Target Gene Details
Entrez Gene ID: | 26900 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 3, Y-linked |
Gene Aliases: |
8030469F12Rik, D1Pas1-rs1, Dby |
Location: |
Chr.Y:1260715-1286628 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ddx3y | NM_012008.2 | 13 | 1457 | NP_036138.1 |
AJ007376.1 | 13 | 1424 | CAA07483.1 | |
AK154382.1 | 13 | 1417 | BAE32549.1 | |
BC021453.1 | 13 | 1422 | AAH21453.1 | |
L25337.1 | 5 | 522 | AAA53631.1 |
