Assay Details
Target Gene Details
Entrez Gene ID: | 26900 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 3, Y-linked |
Gene Aliases: |
8030469F12Rik, D1Pas1-rs1, Dby |
Location: |
Chr.Y:1260715-1286628 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ddx3y | NM_012008.2 | NP_036138.1 | ||
AJ007376.1 | CAA07483.1 | |||
AK154382.1 | BAE32549.1 | |||
BC021453.1 | AAH21453.1 | |||
L25337.1 | AAA53631.1 |
