Assay Details
Target Gene Details
Entrez Gene ID: | 26908 |
Gene Name: | eukaryotic translation initiation factor 2, subunit 3, structural gene Y-linked |
Gene Aliases: |
Eif-2gy, Spy, Tfy |
Location: |
Chr.Y:1010578-1046811 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif2s3y | NM_012011.1 | NP_036141.1 | ||
| AJ006584.1 | CAA07097.1 | |||
| AK009448.1 | BAB26294.1 | |||
| AK012186.1 | BAB28086.1 | |||
| AK153974.1 | ||||
| BC043656.1 | AAH43656.1 |
More Information
Set Membership: |
|

Genomic Map