Assay Details
Target Gene Details
Entrez Gene ID: | 22294 |
Gene Name: | ubiquitously expressed transcript |
Gene Aliases: |
0910002B17Rik |
Location: |
Chr.X:20951665-20962035 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Uxt | NM_013840.3 | NP_038868.2 | ||
| AF092738.1 | AAD28699.1 | |||
| AK027917.1 | BAC25665.1 | |||
| AK038495.1 | ||||
| AK086822.1 | ||||
| AK163983.1 | BAE37569.1 | |||
| BC029258.1 | AAH29258.1 | |||
| BC082294.1 | AAH82294.1 |
More Information
Set Membership: |
|

Genomic Map