Assay Details
Target Gene Details
Entrez Gene ID: | 170828 |
Gene Name: | vestigial like 1 homolog (Drosophila) |
Gene Aliases: |
Tondu |
Location: |
Chr.X:57088079-57106539 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vgll1 | NM_133251.1 | NP_573514.1 | ||
| AB036708.1 | BAB40534.1 |
More Information
Set Membership: |
|

Genomic Map