Assay Details
Target Gene Details
Entrez Gene ID: | 76130 |
Gene Name: | LAS1-like (S. cerevisiae) |
Gene Aliases: |
1810030A06Rik, 5830482G23Rik |
Location: |
Chr.X:95920045-95956974 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Las1l |
Target Gene Details
Entrez Gene ID: | 547176 |
Gene Name: | zinc finger CCCH-type containing 12B |
Gene Aliases: |
A130028J20Rik, CXorf32, Gm373 |
Location: |
Chr.X:95711529-95932656 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zc3h12b | NM_001034907.2 | 6 | 1531 | NP_001030079.2 |
| AK134628.1 | 6 | 1532 | BAE22215.1 |
More Information
Set Membership: |
|

Genomic Map