Assay Details
Target Gene Details
Entrez Gene ID: | 278180 |
Gene Name: | V-set and immunoglobulin domain containing 4 |
Gene Aliases: |
A530061A11, BC025105, CRIg, Z39IG |
Location: |
Chr.X:96247201-96293518 on Build GRCm38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vsig4 | NM_177789.4 | NP_808457.1 | ||
| AK041014.1 | BAC30780.1 | |||
| BC025105.1 | AAH25105.1 |
More Information
Set Membership: |
|

Genomic Map