Assay Details
Target Gene Details
Entrez Gene ID: | 209446 |
Gene Name: | transcription factor E3 |
Gene Aliases: |
F830016E06Rik, Tcfe3, Tfe-3, bHLHe33 |
Location: |
Chr.X:7762538-7775202 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tfe3 | NM_001105196.1 | NP_001098666.1 | ||
NM_001105197.1 | NP_001098667.1 | |||
NM_001271489.1 | NP_001258418.1 | |||
NM_001271490.1 | NP_001258419.1 | |||
NM_001271491.1 | NP_001258420.1 | |||
NM_172472.3 | NP_766060.2 | |||
AK089762.1 | BAC40955.1 | |||
AK139756.1 | BAE24128.1 | |||
AK144657.1 | BAE25994.1 | |||
AK144751.1 | ||||
AK145029.1 | ||||
AK145832.1 | ||||
AK147405.1 | 5 | 1716 | ||
AK149546.1 | 1 | 149 | ||
AK150372.1 | ||||
AK151447.1 | ||||
AK152221.1 | BAE31048.1 | |||
AK153051.1 | ||||
AK154690.1 | ||||
AK155883.1 | BAE33481.1 | |||
AK159162.1 | ||||
AK220116.1 | ||||
BC056358.1 | AAH56358.1 | |||
BC063047.1 | AAH63047.1 | |||
S76673.1 | AAB21130.1 |
