Assay Details
Target Gene Details
Entrez Gene ID: | 26987 |
Gene Name: | eukaryotic translation initiation factor 4E member 2 |
Gene Aliases: |
2700069E09Rik, AI036339, AV129531, D0H0S6743E, Eif4el3 |
Location: |
Chr.1:87213910-87240488 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif4e2 | NM_023314.3 | 7 | 1192 | NP_075803.2 |
| AK018516.1 | 7 | 1190 | BAB31251.1 | |
| AK076067.1 | 5 | 907 | ||
| AK210650.1 | 1 | 52 |
More Information
Set Membership: |
|

Genomic Map