Assay Details
Target Gene Details
Entrez Gene ID: | 226982 |
Gene Name: | eukaryotic translation initiation factor 5B |
Gene Aliases: |
A030003E17Rik, BC018347, IF2 |
Location: |
Chr.1:37998000-38055579 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif5b | NM_198303.2 | 1 | 213 | NP_938045.2 |
| AK145146.1 | 1 | 100 | BAE26259.1 | |
| AK145732.1 | 1 | 214 | BAE26614.1 | |
| AK163527.1 | 1 | 117 | BAE37384.1 | |
| AK168377.1 | 1 | 93 | ||
| BC018347.1 | ||||
| BC037150.1 | AAH37150.1 | |||
| BC040746.1 | 1 | 84 | AAH40746.1 | |
| BC060288.1 | AAH60288.1 |
More Information
Set Membership: |
|

Genomic Map