Assay Details
Target Gene Details
Entrez Gene ID: | 226982 |
Gene Name: | eukaryotic translation initiation factor 5B |
Gene Aliases: |
A030003E17Rik, BC018347, IF2 |
Location: |
Chr.1:37998000-38055579 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif5b | NM_198303.2 | 10 | 1963 | NP_938045.2 |
| AK145146.1 | 10 | 1850 | BAE26259.1 | |
| CN670500.1 | ||||
| CN724918.1 | 5 | 483 |
More Information
Set Membership: |
|

Genomic Map