Assay Details
Target Gene Details
Entrez Gene ID: | 69821 |
Gene Name: | mitochondrial transcription termination factor 4 |
Gene Aliases: |
1810059A23Rik, 4933412H03Rik, Mterfd2 |
Location: |
Chr.1:93299211-93305965 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mterf4 | NM_178051.3 | 2 | 383 | NP_835152.1 |
AK007901.1 | 2 | 370 | ||
AK033043.1 | 2 | 394 | ||
AK077142.1 | 2 | 383 | BAC36640.1 | |
AK079098.1 | 2 | 377 | ||
BC002280.1 | AAH02280.1 | |||
BC055877.1 | 1 | 327 | AAH55877.1 | |
BC064709.1 | 2 | 376 | AAH64709.1 | |
BY261356.1 | 2 | 388 |
