Assay Details
Target Gene Details
Entrez Gene ID: | 74229 |
Gene Name: | progestin and adipoQ receptor family member VIII |
Gene Aliases: |
1700019B16Rik, 3110001D06Rik |
Location: |
Chr.1:20890622-20939650 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Paqr8 | NM_028829.3 | 3 | 941 | NP_083105.3 |
| AF313617.1 | 4 | 1000 | AAO47230.1 | |
| AK006107.2 | 3 | 927 | BAB24412.1 | |
| AK029723.1 | 3 | 922 | ||
| AK029772.1 | 3 | 958 | BAC26609.1 | |
| AK031969.1 | 3 | 954 | BAC27629.1 | |
| AK048045.1 | 3 | 956 | BAC33221.1 | |
| AK147376.1 | 3 | 946 | BAE27872.1 | |
| AY424297.1 | 3 | 956 | AAR08385.1 | |
| BC059813.1 | 3 | 941 | AAH59813.1 |
More Information
Set Membership: |
|

Genomic Map