Assay Details
Target Gene Details
Entrez Gene ID: | 12043 |
Gene Name: | B cell leukemia/lymphoma 2 |
Gene Aliases: |
AW986256, Bcl-2, C430015F12Rik, D630044D05Rik, D830018M01Rik |
Location: |
Chr.1:106538178-106714290 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bcl2 | NM_009741.4 | NP_033871.2 | ||
| NM_177410.2 | 1 | 2111 | NP_803129.2 | |
| AK049473.1 | BAC33767.1 | |||
| AK077913.1 | 1 | 889 | ||
| BC068988.1 | AAH68988.1 | |||
| BC095964.1 | AAH95964.1 |

Genomic Map