Assay Details
Target Gene Details
Entrez Gene ID: | 51810 |
Gene Name: | heterogeneous nuclear ribonucleoprotein U |
Gene Aliases: |
AA408410, AI256620, AL024194, AL024437, AW557595, C86794, Hnrpu, SAFA, Sp120, hnRNP U |
Location: |
Chr.1:178321108-178337810 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hnrnpu | NM_016805.2 | 14 | 2746 | NP_058085.2 |
AK029944.1 | ||||
AK049375.1 | ||||
AK080656.1 | 1 | 36 | ||
AK089058.1 | 14 | 2747 | BAC40725.1 | |
AK135588.1 | 2 | 247 | ||
AK145499.1 | 14 | 2749 | BAE26473.1 | |
AK145682.1 | 14 | 2749 | ||
AK159084.1 | 14 | 2715 | BAE34802.1 | |
AK159614.1 | 14 | 2751 | BAE35231.1 | |
AK159956.1 | 14 | 2688 | BAE35512.1 | |
AK165844.1 | 14 | 2746 | BAE38409.1 | |
AK167200.1 | 14 | 2749 | ||
AK168657.1 | 14 | 2688 | BAE40513.1 | |
BC018353.1 | 14 | 2591 | AAH18353.1 |
