Assay Details
Target Gene Details
Entrez Gene ID: | 70750 |
Gene Name: | 3-ketodihydrosphingosine reductase |
Gene Aliases: |
6330410P18Rik, 9430079B08Rik, Fvt1 |
Location: |
Chr.1:106720410-106759830 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kdsr | NM_027534.2 | 10 | 1739 | NP_081810.1 |
| AK018155.1 | 11 | 1714 | ||
| AK170064.1 | 10 | 1622 | BAE41541.1 | |
| AK219672.1 | 1 | 196 | ||
| BC023820.1 | 10 | 1729 |
More Information
Set Membership: |
|

Genomic Map