Assay Details
Target Gene Details
Entrez Gene ID: | 269224 |
Gene Name: | PAS domain containing serine/threonine kinase |
Gene Aliases: |
mKIAA0135 |
Location: |
Chr.1:93309437-93343350 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pask | NM_080850.2 | 1 | 95 | NP_543126.2 |
| AK028435.1 | 1 | 97 | BAC25951.1 | |
| AK083528.1 | 1 | 66 | ||
| AK083590.1 | 1 | 60 | ||
| AK129063.1 | BAC97873.1 | |||
| AK172031.1 | ||||
| BY058767.1 | 1 | 95 |
More Information
Set Membership: |
|

Genomic Map