Assay Details
Target Gene Details
Entrez Gene ID: | 54354 |
Gene Name: | Ras association (RalGDS/AF-6) domain family member 5 |
Gene Aliases: |
1300019G20Rik, AU042887, Maxp1, Nore1, Nore1A, Nore1B, Rapl |
Location: |
Chr.1:131176410-131245178 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rassf5 | NM_018750.3 | 1 | 331 | NP_061220.2 |
| AF053959.1 | 1 | 313 | AAC08580.1 | |
| AK005059.2 | 1 | 327 | ||
| AK029321.1 | 1 | 332 | ||
| AK155534.1 | 1 | 327 | BAE33312.1 | |
| BC089605.1 | 1 | 333 | AAH89605.1 |
More Information
Set Membership: |
|

Genomic Map