Assay Details
Target Gene Details
Entrez Gene ID: | 16865 |
Gene Name: | eukaryotic translation initiation factor 2D |
Gene Aliases: |
D1Ertd5e, Lgtn |
Location: |
Chr.1:131153170-131187659 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif2d | AK030312.1 | |||
| AK036154.1 | BAC29323.1 | |||
| AK047005.1 | BAC32939.1 |
Target Gene Details
Entrez Gene ID: | 54354 |
Gene Name: | Ras association (RalGDS/AF-6) domain family member 5 |
Gene Aliases: |
1300019G20Rik, AU042887, Maxp1, Nore1, Nore1A, Nore1B, Rapl |
Location: |
Chr.1:131176410-131245178 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rassf5 | NM_018750.3 | 6 | 1898 | NP_061220.2 |
| AF053959.1 | 6 | 1880 | AAC08580.1 | |
| AK005059.2 | 6 | 1894 | ||
| AK029321.1 | 6 | 1803 | ||
| AK088751.1 | 5 | 1533 | BAC40546.1 | |
| AK131147.1 | 5 | 1491 | BAD21397.1 | |
| AK155534.1 | 6 | 1894 | BAE33312.1 | |
| AK155869.1 | 5 | 1570 | BAE33472.1 | |
| BC089605.1 | 6 | 1900 | AAH89605.1 |
More Information
Set Membership: |
|

Genomic Map