Assay Details
Target Gene Details
Entrez Gene ID: | 74205 |
Gene Name: | acyl-CoA synthetase long-chain family member 3 |
Gene Aliases: |
2610510B12Rik, Acs3, C85929, Facl3, Pro2194 |
Location: |
Chr.1:78657825-78707743 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acsl3 | NM_001033606.2 | 15 | 3560 | NP_001028778.2 |
| NM_001136222.1 | 14 | 3142 | NP_001129694.1 | |
| NM_028817.3 | 16 | 3700 | NP_083093.2 | |
| AK046000.1 | 1 | 1007 | ||
| CK379747.1 | 1 | 208 |
More Information
Set Membership: |
|

Genomic Map