Assay Details
Target Gene Details
Entrez Gene ID: | 51800 |
Gene Name: | BCL2-related ovarian killer |
Gene Aliases: |
AI847676, AU021146, matador, mtd |
Location: |
Chr.1:93685694-93695762 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bok | NM_016778.2 | 5 | 1072 | NP_058058.1 |
AF027707.1 | 5 | 1070 | AAC53582.1 | |
AK156243.1 | 5 | 1107 | BAE33639.1 | |
AK168373.1 | 5 | 1003 | BAE40305.1 | |
AK171258.1 | 5 | 1029 | BAE42349.1 | |
BC030069.1 | 5 | 1010 | AAH30069.1 |
