Assay Details
Target Gene Details
Entrez Gene ID: | 15528 |
Gene Name: | heat shock protein 1 (chaperonin 10) |
Gene Aliases: |
10kDa, Hsp10, mt-cpn10 |
Location: |
Chr.1:55088148-55091317 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hspe1 | NM_008303.4 | 1 | 43 | NP_032329.1 |
| AV570511.1 | 1 | 43 |
More Information
Set Membership: |
|

Genomic Map