Assay Details
Target Gene Details
Entrez Gene ID: | 18566 |
Gene Name: | programmed cell death 1 |
Gene Aliases: |
Ly101, PD-1, Pdc1 |
Location: |
Chr.1:94038305-94052553 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pdcd1 | NM_008798.2 | 5 | 1726 | NP_032824.1 |
| AK039828.1 | 5 | 1706 | BAC30463.1 | |
| X67914.1 | 5 | 1726 | CAA48113.1 |

Genomic Map