Assay Details
Target Gene Details
Entrez Gene ID: | 271639 |
Gene Name: | adenylate cyclase 10 |
Gene Aliases: |
4930431D04Rik, 4931412F17, Sacy, sAC |
Location: |
Chr.1:165483869-165576778 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Adcy10 | NM_173029.3 | 21 | 2980 | NP_766617.2 |
AK029849.1 | 21 | 2982 | BAC26642.1 | |
AK133181.1 | 20 | 2868 | BAE21546.1 | |
BC138448.1 | 21 | 2861 |
