Assay Details
Target Gene Details
Entrez Gene ID: | 107508 |
Gene Name: | glutamyl-prolyl-tRNA synthetase |
Gene Aliases: |
2410081F06Rik, 3010002K18Rik, C79379, Qprs |
Location: |
Chr.1:185363028-185428355 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eprs | NM_029735.1 | 18 | 2401 | NP_084011.1 |
| AK028687.1 | 11 | 1422 | ||
| AK044884.1 | 1 | 1768 | ||
| AK141072.1 | 18 | 2456 | ||
| AK148463.1 | 18 | 2362 | BAE28568.1 | |
| AK160986.1 | 18 | 2421 | ||
| BC040802.1 | 18 | 2399 | AAH40802.1 | |
| BC094679.1 | 17 | 2195 | AAH94679.1 | |
| BC141049.1 | 18 | 2320 | ||
| X54327.1 | 9 | 1085 |
More Information
Set Membership: |
|

Genomic Map