Assay Details
Target Gene Details
Entrez Gene ID: | 22592 |
Gene Name: | excision repair cross-complementing rodent repair deficiency, complementation group 5 |
Gene Aliases: |
Xpg |
Location: |
Chr.1:44147578-44181260 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ercc5 | NM_011729.2 | 15 | 3403 | NP_035859.2 |
| AK137563.1 | 16 | 3235 | BAE23409.1 | |
| AK140825.1 | ||||
| AK164205.1 | 15 | 3221 | BAE37682.1 | |
| BC060659.1 | 14 | 3534 | AAH60659.1 | |
| BC141070.1 | 15 | 3233 | ||
| D16306.1 | 15 | 3098 | BAA03813.1 | |
| U40796.1 | 15 | 3299 | AAA91039.1 |
More Information
Set Membership: |
|

Genomic Map