Assay Details
Target Gene Details
Entrez Gene ID: | 213464 |
Gene Name: | retinoblastoma binding protein 5 |
Gene Aliases: |
4933411J24Rik, C330016J05 |
Location: |
Chr.1:132477347-132505665 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rbbp5 | NM_172517.2 | 14 | 3000 | NP_766105.2 |
| AK016773.1 | 1 | 1301 | BAB30420.1 | |
| BC057632.1 | 15 | 2759 | AAH57632.1 | |
| BC060186.1 | 3 | 1443 |

Genomic Map